2'-Methoxyacetophenone, 98%
2'-Methoxyacetophenone, 98%
2'-Methoxyacetophenone, 98%
2'-Methoxyacetophenone, 98%
Thermo Scientific Chemicals
2'-Methoxyacetophenone, 98%
CAS: 579-74-8 | C9H10O2 | 150.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11929.1425 g
Catalog number ALFA11929.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2'-Methoxyacetophenone
CAS579-74-8
Health Hazard 1H302-H315-H319
Health Hazard 3P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c