Isopropyl cyanoacetate, 97%
Isopropyl cyanoacetate, 97%
Isopropyl cyanoacetate, 97%
Isopropyl cyanoacetate, 97%
Thermo Scientific Chemicals
Isopropyl cyanoacetate, 97%
CAS: 13361-30-3 | C6H9NO2 | 127.143 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA11887.22
Price (MYR)
622.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
622.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsopropyl cyanoacetate
CAS13361-30-3
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing KnÖevenagel reactions with aldehydes and ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing Knöevenagel reactions with aldehydes and ketones.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Kohei Yamashita; Takanori Tanaka; Masahiko Hayashi. Use of isopropyl alcohol as a solvent in Ti(O-i-Pr)4-catalyzed Knöevenagel reactions. Tetrahedron,2005, 61(33), 7981-7985.
  2. Chan Hyuk Kim; Ki Po Jang; Soo Young Choi; Young Keun Chung; and Eun Lee . A Carbonyl Ylide Cycloaddition Approach to Platensimycin. Angewandte Chemie International Edition,2008, 47(47), 4009-4011.