Theobromine, 99%
Theobromine, 99%
Theobromine, 99%
Thermo Scientific Chemicals
Theobromine, 99%
Phosphodiesterase inhibitor | CAS: 83-67-0 | C7H8N4O2 | 180.167 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11861.1850 g
Catalog number ALFA11861.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTheobromine
CAS83-67-0
Health Hazard 1H302
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
Phosphodiesterase inhibitor, weak adenosine receptor antagonist, diuretic and smooth muscle relaxant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphodiesterase inhibitor, weak adenosine receptor antagonist, diuretic and smooth muscle relaxant.
Solubility
Soluble in hot methanol and dimethyl sulfoxide.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store away from strong bases.
RUO – Research Use Only
General References:
  1. Monika Johannsen; Gerd Brunner. Solubilities of the xanthines caffeine, theophylline and theobromine in supercritical carbon dioxide. Fluid Phase Equilibria.,1994, 95(1), 215-226.
  2. Kreiser WR; Martin RA Jr. High pressure liquid chromatographic determination of theobromine and caffeine in cocoa and chocolate products. Journal - Association of Official Analytical Chemists,1978, 61(6), 1424-1427.