4,4'-Stilbenedicarboxylic acid, 96%
4,4'-Stilbenedicarboxylic acid, 96%
4,4'-Stilbenedicarboxylic acid, 96%
4,4'-Stilbenedicarboxylic acid, 96%
4,4'-Stilbenedicarboxylic acid, 96%
CAS: 100-31-2 | C16H12O4 | 268.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Catalog number ALFA11839.14
Price (MYR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4,4'-Stilbenedicarboxylic acid
CAS100-31-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c