Glycerol tributyrate, 97%
Glycerol tributyrate, 97%
Glycerol tributyrate, 97%
Glycerol tributyrate, 97%
Thermo Scientific Chemicals
Glycerol tributyrate, 97%
Glycerol tributyrate, 98%, C15H26O6, CAS Number-60-01-5, tri-n-butyrin, glyceryl tributyrate, propane-1,2,3-triyl tributyrate, glycerol tributyrate, glycerol tributanoate, glyceroltributyrin, tributyrin, butyrin, butyryl triglyceride, tributin, 1000g, 1714746, 307 deg.C to 308 deg.C, CHEBI:35020 | CAS: 60-01-5 | C15H26O6 | 302.367 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11830.30250 g
Catalog number ALFA11830.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycerol tributyrate
CAS60-01-5
Melting Point-75°C
Recommended StorageAmbient temperatures
Density1.032
View more
Glycerol tributyrate is a triglyceride that may inhibit cell growth and induce cell differentiation. It is used to to induce differentiation of malignant cells followed by arrest of cell growth and apoptosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycerol tributyrate is a triglyceride that may inhibit cell growth and induce cell differentiation. It is used to to induce differentiation of malignant cells followed by arrest of cell growth and apoptosis.
Solubility
Not miscible in water.
Notes
Incompatible with strong oxidizing agents, Strong acids, Strong bases. Use with adequate ventilation.
RUO – Research Use Only
Nack-Shick Choi; Jong Hyun Choi; Bo-Hye Kim; Yun-Jon Han; Joong Su Kim; Seung-Goo Lee and Jae Jun Song. Mixed-substrate (glycerol tributyrate and fibrin) zymography for simultaneous detection of lipolytic and proteolytic enzymes on a single gel.Electrophoresis.2009, 30 2234-2237.