2-Methylresorcinol, 98%
2-Methylresorcinol, 98%
2-Methylresorcinol, 98%
2-Methylresorcinol, 98%
Thermo Scientific Chemicals
2-Methylresorcinol, 98%
CAS: 608-25-3 | C7H8O2 | 124.139 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11821.1850 g
Catalog number ALFA11821.18
Price (MYR)
383.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
383.00
EA
Add to cart
Specifications
Chemical Name or Material2-Methylresorcinol
CAS608-25-3
Health Hazard 1H301-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
2-Methylresorcinol is used in the preparation of aromatic benziporphyrins, tripyrrane analogs and C-5-bromo-2-hydroxyphenylcalix[4]-2-methylresorcinarene. Further, it is employed in oxidative and non-oxidative hair dye formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methylresorcinol is used in the preparation of aromatic benziporphyrins, tripyrrane analogs and C-5-bromo-2-hydroxyphenylcalix[4]-2-methylresorcinarene. Further, it is employed in oxidative and non-oxidative hair dye formulations.
Solubility
Soluble in water and methanol.
Notes
Incompatible with acid chlorides, acid anhydrides and oxidizing agents
RUO – Research Use Only
General References:
  1. Mir, N. A.; Dubey, R.; Tothadi, S.; Desiraju, G. R. Combinatorial crystal synthesis of ternary solids based on 2-methylresorcinol. CrystEngComm 2015, 17 (41), 7866-7869.
  2. Oie, H.; Mori, A.; Sudo, A.; Endo, T. Polyaddition of bifunctional 1,3-benzoxazine and 2-methylresorcinol. J. Polym. Sci., Part A: Polym. Chem. 2013, 51 (18), 3867-3872.