Sodium n-octyl sulfate, 99%
Sodium n-octyl sulfate, 99%
Sodium n-octyl sulfate, 99%
Sodium n-octyl sulfate, 99%
Thermo Scientific Chemicals
Sodium n-octyl sulfate, 99%
CAS: 142-31-4 | C8H17NaO4S | 232.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA11786.14
Price (MYR)
2,318.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,318.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium n-octyl sulfate
CAS142-31-4
Health Hazard 1H315-H318
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
View more
Sodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Lu, J.; Ni, X.; Cao, Y.; Ma, X.; Cao, G. Electrokinetic chromatographic characterization of novel catanionic surfactants vesicle as pseudostationary phase. Electrophoresis 2015, 36 (2), 312-318.
  2. Bijttebier, S.; D’Hondt, E.; Noten, B.; Hermans, N.; Apers, S.; Voorspoels, S. Improving method reliability in carotenoid analysis through selective removal of glycerolipid interferences by lipase treatment. J. Agric. Food. Chem. 2014, 62 (14), 3114-3124.