Lead(II) acetate trihydrate, 99%
Lead(II) acetate trihydrate, 99%
Lead(II) acetate trihydrate, 99%
Lead(II) acetate trihydrate, 99%
Thermo Scientific Chemicals
Lead(II) acetate trihydrate, 99%
CAS: 6080-56-4 | C4H12O7Pb | 379.30 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11746.30250 g
Catalog number ALFA11746.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLead (II) acetate trihydrate
CAS6080-56-4
Health Hazard 1H360D-H361f-H372
Health Hazard 2GHS H Statement
H360-H373
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
View more
Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.
Solubility
Freely soluble in water, alcohol and glycerol.
Notes
Decomposes above 200°Air and light sensitive. Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Knichal, J. V.; Gee, W. J.; Burrows, A. D.; Raithby, P. R.; Wilson, C. C. A new small molecule gelator and 3D framework ligator of lead(II). CrystEngComm 2015, 17 (42), 8139-8145.
  2. Sun, X.; Yang, J.; Zhang, W.; Zhu, X.; Hu, Y.; Yang, D.; Yuan, X.; Yu, W.; Dong, J.; Wang, H.; Li, L.; Kumar, V. R.; Liang, S. Lead acetate trihydrate precursor route to synthesize novel ultrafine lead oxide from spent lead acid battery pastes. J. Power Sources 2014, 269, 565-576.