2-Mercaptopyridine, 98%
2-Mercaptopyridine, 98%
2-Mercaptopyridine, 98%
2-Mercaptopyridine, 98%
Thermo Scientific Chemicals
2-Mercaptopyridine, 98%
CAS: 2637-34-5 | C5H5NS | 111.162 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11741.065 g
Catalog number ALFA11741.06
Price (MYR)
315.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
315.00
EA
Add to cart
Specifications
Chemical Name or Material2-Mercaptopyridine
CAS2637-34-5
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.
Solubility
Slightly soluble in chloroform, methanol and dimethyl sulfoxide.
Notes
Air Sensitive, keep the container tightly sealed. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. David B Cordes; and Lyall R Hanton. Structural invariance in silver(I) coordination networks formed using flexible four-armed thiopyridine ligands. Inorganic Chemistry. 2007, 46 (5), 1634-1644.
  2. Alexey V Chuchuryukin et. al. Hydroxy- and mercaptopyridine pincer platinum and palladium complexes generated by silver-free halide abstraction. Inorganic Chemistry, 2006, 45 (5), 2045-2054.
  3. S-Acyl derivatives react with Grignard reagents in a high-yielding synthesis of ketones: J. Am. Chem. Soc., 95, 4763 (1973):
  4. Titanium enolates of S-acyl derivatives react with imines in a one pot synthesis of ß-lactams: J. Org. Chem., 57, 4155 (1992).