2,3-Dichloroquinoxaline, 98%
2,3-Dichloroquinoxaline, 98%
2,3-Dichloroquinoxaline, 98%
2,3-Dichloroquinoxaline, 98%
Thermo Scientific Chemicals
2,3-Dichloroquinoxaline, 98%
CAS: 2213-63-0 | C8H4Cl2N2 | 199.034 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11734.065 g
Catalog number ALFA11734.06
Price (MYR)
337.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
337.00
EA
Add to cart
Specifications
Chemical Name or Material2,3-Dichloroquinoxaline
CAS2213-63-0
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.