3'-Fluoroacetophenone, 97%
3'-Fluoroacetophenone, 97%
3'-Fluoroacetophenone, 97%
3'-Fluoroacetophenone, 97%
Thermo Scientific Chemicals
3'-Fluoroacetophenone, 97%
CAS: 455-36-7 | C8H7FO | 138.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11733.065 g
Catalog number ALFA11733.06
Price (MYR)
381.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
381.00
EA
Add to cart
Specifications
Chemical Name or Material3'-Fluoroacetophenone
CAS455-36-7
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
3'-Fluoroacetophenone is used as raw material and intermediate in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3′-Fluoroacetophenone is used as raw material and intermediate in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents. Ensure adequate ventilation during use. Use only in a chemical fume hood.
RUO – Research Use Only
General References:
  1. Jan von Langermann; Annett Mell;, Eckhard Paetzold; Thomas Daußmann and Udo Kragl. Hydroxynitrile Lyase in Organic Solvent-Free Systems to Overcome Thermodynamic Limitations.Advanced Synthesis & Catalysis. 2007, 349 1418-1424.
  2. Asgeir Bjarnason; James W. Taylor; James A. Kinsinger; Robert B. Cody; David A. Weil. Isomer discrimination of disubstituted benzene derivatives through gas-phase iron(I) ion reactions in a Fourier-transform mass spectrometer.Anal. Chem. 1989, 61 (17), 1889-1894.