Hydantoin-5-acetic acid, 98%
Hydantoin-5-acetic acid, 98%
Hydantoin-5-acetic acid, 98%
Hydantoin-5-acetic acid, 98%
Thermo Scientific Chemicals
Hydantoin-5-acetic acid, 98%
CAS: 5427-26-9 | C5H6N2O4 | 158.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11731.065 g
Catalog number ALFA11731.06
Price (MYR)
306.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
306.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydantoin-5-acetic acid
CAS5427-26-9
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.