Diphenylphosphinic chloride, 97+%
Diphenylphosphinic chloride, 97+%
Diphenylphosphinic chloride, 97+%
Diphenylphosphinic chloride, 97+%
Thermo Scientific Chemicals
Diphenylphosphinic chloride, 97+%
CAS: 1499-21-4 | C12H10ClOP | 236.63 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11721.22100 g
Catalog number ALFA11721.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDiphenylphosphinic chloride
CAS1499-21-4
Health Hazard 1H226-H314
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.
Solubility
Decomposes with water.
Notes
Moisture Sensitive. Store away from strong bases, moisture. Incompatible with oxidizing agents,, Amines, Alcohols.
RUO – Research Use Only
General References:
  1. K. Darrell Berlin; T. Howard Austin; M. Nagabhushanam. A Convenient Synthesis of Esters of Diphenylphosphinic Acid. J. Org. Chem. 1965, 30 (4), 1267-1268.
  2. Shaoqing Chen; Jiecheng Xu. Pentafluorophenyl diphenylphosphinate a new efficient coupling reagent in peptide chemistry.Tetrahedron letters. 1991, 32 6711-6714.