Boron oxide, 98%
Boron oxide, 98%
Boron oxide, 98%
Thermo Scientific Chemicals
Boron oxide, 98%
CAS: 1303-86-2 | B2O3 | 69.62 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11707.36500 g
Catalog number ALFA11707.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point1860°C
Chemical Name or MaterialBoron oxide
Melting Point∽450°C
CAS1303-86-2
Health Hazard 1H360FD
View more
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.
Solubility
Soluble in water. Slightly soluble in methanol.
Notes
Hygroscopic and moisture sensitive. Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Kline, J.; Tangstad, M.; Tranell, G. A Raman Spectroscopic Study of the Structural Modifications Associated with the Addition of Calcium Oxide and Boron Oxide to Silica. Metall. Mater. Trans. B 2015, 46 (1), 62-73.
  2. Solozhenko, V. L.; Kurakevych, O. O.; Godec, Y. L.; Brazhkin, V. V. Thermodynamically Consistent p-T Phase Diagram of Boron Oxide B2O3 by in Situ Probing and Thermodynamic Analysis. J. Phys. Chem. C 2015, 119 (35), 20600-20605.