2-Chloro-2-methylbutane, 95%
2-Chloro-2-methylbutane, 95%
2-Chloro-2-methylbutane, 95%
2-Chloro-2-methylbutane, 95%
Thermo Scientific Chemicals
2-Chloro-2-methylbutane, 95%
CAS: 594-36-5 | C5H11Cl | 106.593 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11703.1425 g
Catalog number ALFA11703.14
Price (MYR)
646.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
646.00
EA
Add to cart
Specifications
Chemical Name or Material2-Chloro-2-methylbutane
CAS594-36-5
Health Hazard 1H225
Health Hazard 2GHS H Statement
H225
Highly flammable liquid and vapor.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
View more
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.
Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents. Avoid extremes of temperature and direct sunlight.
RUO – Research Use Only
Lídia M.P.C. Albuquerque; M.Luisa C.J. Moita; M.N. Simoes; Raquel M.C. Goncalves. The enthalpies of solution of 2-chloro-2-methylpropane, 2-chloro-2-methylbutane and 3-chloro-3-methylpentane in mono- and dialcohols at 298.15 K. Thermochimica acta. 1996, 275 67-73.