2,4,6-Trichlorophenol, 98%
2,4,6-Trichlorophenol, 98%
2,4,6-Trichlorophenol, 98%
2,4,6-Trichlorophenol, 98%
Thermo Scientific Chemicals
2,4,6-Trichlorophenol, 98%
CAS: 88-06-2 | C6H3Cl3O | 197.439 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1000 g
Catalog number ALFA11681.30
Price (MYR)
469.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
469.00
EA
Add to cart
Specifications
Chemical Name or Material2,4,6-Trichlorophenol, 98%
CAS88-06-2
Health Hazard 1H302-H315-H319-H351
Health Hazard 2GHS H Statement
H351-H302-H315-H319
Health Hazard 3P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
2,4,6-Trichlorophenol, antiseptic, defoliant, and glue preservative, chemical intermediate, anti-mildew treatment, solvent for polyester fiber.
Solubility
Soluble in water(0.8g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Irfan Z. Shirgaonkar, Aniruddha B. Pandit. Sonophotochemical destruction of aqueous solution of 2,4,6-trichlorophenol. Ultrasonics Sonochemistry. 1998, 5, (2), 53-61.
  2. Christopher J. Daughney.; Jeremy B. Fein. Sorption of 2,4,6-Trichlorophenol by Bacillus subtilis. Environ. Sci. Technol. 1998, 32, (6), 749-752.