Calcium D-gluconate monohydrate, 98+%
Calcium D-gluconate monohydrate, 98+%
Calcium D-gluconate monohydrate, 98+%
Calcium D-gluconate monohydrate, 98+%
Thermo Scientific Chemicals
Calcium D-gluconate monohydrate, 98+%
CAS: 66905-23-5 | C12H24CaO15 | 448.387 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA11649.36
Price (MYR)
334.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
334.00
EA
Add to cart
Specifications
Chemical Name or MaterialCalcium D-gluconate monohydrate
CAS66905-23-5
Melting Point∼195°C (decomposition)
Recommended StorageAmbient temperatures
RTECS NumberEW2100000
View more
Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.
Solubility
Soluble in water(30g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Panagiotopoulos,N.C.; Jeffrey, G.A.; La Placa, S.J.; Hamilton, W.C. The crystal structures of the A and B forms of potassium D-gluconate monohydrate by neutron diffraction. Acta Cryst. 1974, 30, 1421-1430.
  2. Martina Vavrusova.; Merete Bogelund Munk.; Leif H. Skibsted. Aqueous Solubility of Calcium l-Lactate, Calcium d-Gluconate, and Calcium d-Lactobionate: Importance of Complex Formation for Solubility Increase by Hydroxycarboxylate Mixture. J. Agric. Food Chem. 2013, 61, (34), 8207-8214.