3-(3,4-Dihydroxyphenyl)propionic acid, 98+%
3-(3,4-Dihydroxyphenyl)propionic acid, 98+%
3-(3,4-Dihydroxyphenyl)propionic acid, 98+%
3-(3,4-Dihydroxyphenyl)propionic acid, 98+%
Thermo Scientific Chemicals
3-(3,4-Dihydroxyphenyl)propionic acid, 98+%
CAS: 1078-61-1 | C9H10O4 | 182.175 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11638.065 g
Catalog number ALFA11638.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3- (3,4-Dihydroxyphenyl)propionic acid
CAS1078-61-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
3-(3,4-Dihydroxyphenyl)propionic acid it can be used to produce 3-(3,4-dihydroxy-phenyl)-propionic acid octyl ester by heating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(3,4-Dihydroxyphenyl)propionic acid it can be used to produce 3-(3,4-dihydroxy-phenyl)-propionic acid octyl ester by heating.
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Annett Mikolasch,.; Elke Hammer.; Ulrike Jonas.; Katrin Popowski.; Anne Stielow.; Frieder Schauer. Synthesis of 3-(3,4-dihydroxyphenyl)-propionic acid derivatives by N-coupling of amines using laccase. Tetrahedron. 2002, 58, (38), 7589-7593.
  2. Selma Saraç.; Bezhan Chankvetadze.; Gottfried Blaschke. Enantioseparation of 3,4-dihydroxyphenylalanine and 2-hydrazino-2-methyl-3-(3,4-dihydroxyphenyl)propanoic acid by capillary electrophoresis using cyclodextrins. Journal of Chromatography A. 2000, 875, (1-2), 379-387.