2,4-Difluorophenylhydrazine hydrochloride, 97%
2,4-Difluorophenylhydrazine hydrochloride, 97%
2,4-Difluorophenylhydrazine hydrochloride, 97%
2,4-Difluorophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals
2,4-Difluorophenylhydrazine hydrochloride, 97%
CAS: 51523-79-6 | C6H6F2N2 | 144.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11625.1425 g
Catalog number ALFA11625.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2,4-Difluorophenylhydrazine hydrochloride
CAS51523-79-6
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c