Bromocyclohexane, 98%
Bromocyclohexane, 98%
Bromocyclohexane, 98%
Bromocyclohexane, 98%
Thermo Scientific Chemicals
Bromocyclohexane, 98%
CAS: 108-85-0 | C6H11Br | 163.06 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11593.22100 g
Catalog number ALFA11593.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBromocyclohexane
CAS108-85-0
Health Hazard 1H227
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P280-P370+P378q-P501c
View more
Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air & Light Sensitive. Store away from strong bases.
RUO – Research Use Only
General References:
  1. Songhee Han; Hyun Sik Yoo; Sang Kyu Kim. Conformer-specific ionization spectroscopy of bromocyclohexane: equatorial versus axial conformers. Journal of Physical Chemistry A: Molecules, Spectroscopy, Kinetics, Environment and General Theory.2010, 114 (37), 10005-10010.
  2. Gagan Deep; Rahul Kumar; Anil K Jain; Chapla Agarwal; Rajesh Agarwal. Isolation of axial conformers of chloro- and bromocyclohexane in a pure state as inclusion complexes with 9,9'-bianthryl, and the discovery of a novel 1,3 diaxial Cl...H weak interaction..Chemical Communications.2005, (29), 3646-3648.