2-Fluorobenzyl chloride, 98+%
2-Fluorobenzyl chloride, 98+%
2-Fluorobenzyl chloride, 98+%
2-Fluorobenzyl chloride, 98+%
Thermo Scientific Chemicals
2-Fluorobenzyl chloride, 98+%
CAS: 345-35-7 | C7H6ClF | 144.57 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11566.1425 g
Catalog number ALFA11566.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Fluorobenzyl chloride
CAS345-35-7
Health Hazard 1H226-H302+H312+H332-H314
Health Hazard 2GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c