3,4-Dihydroxybenzaldehyde, 98%
3,4-Dihydroxybenzaldehyde, 98%
3,4-Dihydroxybenzaldehyde, 98%
3,4-Dihydroxybenzaldehyde, 98%
Thermo Scientific Chemicals
3,4-Dihydroxybenzaldehyde, 98%
CAS: 139-85-5 | C7H6O3 | 138.122 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11558.1425 g
Catalog number ALFA11558.14
Price (MYR)
1,123.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,123.00
EA
Add to cart
Specifications
Chemical Name or Material3,4-Dihydroxybenzaldehyde
CAS139-85-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.
Solubility
Soluble in water(6.3g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Nematollahia, D.; Golabi, S.M. Investigation of the electro-methoxylation reaction: Part 1. Electrochemical study of 4-tert-butylcatechol and 3,4-dihydroxybenzaldehyde in methanol. Journal of Electroanalytical Chemistry. 2000, 481, (2), 208-214.
  2. Pariente, F.; Lorenzo, E.; Abruna, H.D. Electrocatalysis of NADH Oxidation with Electropolymerized Films of 3,4-Dihydroxybenzaldehyde. Anal. Chem. 1994, 66, (23), 4337-4344.