Molecular sieves, 4A, powder
Molecular sieves, 4A, powder
Molecular sieves, 4A, powder
Molecular sieves, 4A, powder
Thermo Scientific Chemicals
Molecular sieves, 4A, powder
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11535.30250 g
Catalog number ALFA11535.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMolecular sieves 4A
SensitivityHygroscopic
Quantity250 g
CAS70955-01-0
Physical FormPowder
View more
Molecular sieves, 4 Å is used to absorb molecules as large as butane. It is utilized for the separation of natural gas, liquified petroleum gas (LPG) and alkenes. It finds application in the refining and purification of liquid and gases. It is also used for drying and purification of air, inert gases and solvents. It is actively involved to remove methanol, carbon dioxide and ammonia from fluid streams. It plays a vital role to remove heavy metal ions available in water. It is also used as a soap forming agent and as a detergent auxiliary. It has been considered as a general-purpose drying agent in polar and non polar media.