Guanine hydrochloride, 98%, Thermo Scientific Chemicals
Guanine hydrochloride, 98%, Thermo Scientific Chemicals
Guanine hydrochloride, 98%, Thermo Scientific Chemicals
Guanine hydrochloride, 98%, Thermo Scientific Chemicals
Guanine hydrochloride, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA11532.0910 g
Catalog number ALFA11532.09
Price (MYR)
-
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGuanine hydrochloride
CAS635-39-2
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Guanine hydrochloride is one of the strongest denaturant. It is mainly used in protein purification and protein folding studies. It is a nucleobase used in biochemical research.
Solubility
Slightly soluble in water
Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Sadleir, K. R.; Eimer, W. A.; Cole, S. L.; Vassar, R. A β reduction in BACE1 heterozygous null 5XFAD mice is associated with transgenic APP level. Mol. Neurodegener. 2015, 10 (1), 1-16.
  2. Sahoo, D. K.; Sahu, P. K. Stability-Indicating RP-HPLC Method for the Determination of Valaciclovir Hydrochloride in Bulk and Pharmaceutical Dosage Forms. Chem. Sci. Trans. 2014, 3 (2), 510-517.