Tetra-n-propylammonium bromide, 98%
Tetra-n-propylammonium bromide, 98%
Tetra-n-propylammonium bromide, 98%
Tetra-n-propylammonium bromide, 98%
Thermo Scientific Chemicals
Tetra-n-propylammonium bromide, 98%
CAS: 1941-30-6 | C12H28BrN | 266.267 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11522.22100 g
Catalog number ALFA11522.22
Price (MYR)
405.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
405.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetra-n-propylammonium bromide
CAS1941-30-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Tetra-n-propylammonium bromide, is used as surface-active agents, Solvents, Intermediates, Active Ingredient for Conditioners, Antistatic Agent, Detergent Sanitisers, Softner for textiles and paper products, Phase Transfer Catalyst, Antimicrobials, Disinfection Agents And Sanitizers, Slimicidal Agents, Emulsifying Agents and Pigment Dispersers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetra-n-propylammonium bromide, is used as surface-active agents, Solvents, Intermediates, Active Ingredient for Conditioners, Antistatic Agent, Detergent Sanitisers, Softner for textiles and paper products, Phase Transfer Catalyst, Antimicrobials, Disinfection Agents And Sanitizers, Slimicidal Agents, Emulsifying Agents and Pigment Dispersers.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to Strong oxidizing agents. Stable under ordinary conditions (Hygroscopic)
RUO – Research Use Only
General References:
  1. Kyrre Thalberg.; Bjoern Lindman. Interaction between hyaluronan and cationic surfactants. J. Phys. Chem. 1989, 93, (4), 1478-1483.
  2. Herbert O. House.; Edith Feng.; Norton P. Peet. Comparison of various tetraalkylammonium salts as supporting electrolytes in organic electrochemical reactions. J. Org. Chem. 1971, 36, (16), 2371-2375.