Hexafluorobenzene, 99%
Hexafluorobenzene, 99%
Hexafluorobenzene, 99%
Hexafluorobenzene, 99%
Thermo Scientific Chemicals
Hexafluorobenzene, 99%
CAS: 392-56-3 | C6F6 | 186.056 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11500.0910 g
Catalog number ALFA11500.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHexafluorobenzene
CAS392-56-3
Health Hazard 1H225
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
Melting Point4°C to 5°C
View more
Hexafluorobenzene is used as a solvent in photochemical reactions. It is also used as a reference compound in fluorine-19 NMR, carbon-13 NMR. It is used as a solvent in proton NMR, IR spectrum and UV-spectra. It is used as anticorrosive, antifriction and anti-tumor agents. Further, it is used as a reference molecule to investigate tissue oxygenation studies. It forms series of 1:1 complexes with naphthalene, anthracene, phenanthrene, pyrene and triphenylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexafluorobenzene is used as a solvent in photochemical reactions. It is also used as a reference compound in fluorine-19 NMR, carbon-13 NMR. It is used as a solvent in proton NMR, IR spectrum and UV-spectra. It is used as anticorrosive, antifriction and anti-tumor agents. It forms series of 1:1 complexes with naphthalene, anthracene, phenanthrene, pyrene and triphenylene.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Exchanges with Grignard reagents, under catalysis by CoCl2, to give the pentafluorophenyl Grignard reagent: J. Organomet. Chem., 19, 191 (1969).
  2. Reacts with ethyl cyanoacetate, in the presence of K2CO3 in DMF, to displace one F atom, as the first step of a synthesis of pentafluorophenylacetonitrile: Org. Synth. Coll., 6, 873 (1988).
  3. Gong, S.; Luo, Q.; Feng, X.; Li, Q.-s.; Xie, Y.; King, R. B.; Schaefer Iii, H. F. Triple decker sandwiches and related compounds of the first row transition metals with cyclopentadienyl and hexafluorobenzene rings: remarkable effects of fluorine substitution. Phys. Chem. Chem. Phys. 2015, 17 (31), 20100-20113.
  4. Ma, W. C.; Tsai, C. Y.; Huang, C. Investigation of atmospheric-pressure plasma deposited hexafluorobenzene fluorocarbon film. Surf. Coat. Technol. 2014, 259, 290-296.