2-Amino-5-mercapto-1,3,4-thiadiazole, 98%
2-Amino-5-mercapto-1,3,4-thiadiazole, 98%
2-Amino-5-mercapto-1,3,4-thiadiazole, 98%
2-Amino-5-mercapto-1,3,4-thiadiazole, 98%
Thermo Scientific Chemicals
2-Amino-5-mercapto-1,3,4-thiadiazole, 98%
CAS: 2349-67-9 | C2H3N3S2 | 133.187 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA11451.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-5-mercapto-1,3,4-thiadiazole
CAS2349-67-9
Health Hazard 1H315-H320-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c