Specifications
Chemical Name or Material1,1,3,3-Tetramethoxypropane
CAS102-52-3
Health Hazard 1H226
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
View more