3-Fluorobenzaldehyde, 97%
3-Fluorobenzaldehyde, 97%
3-Fluorobenzaldehyde, 97%
3-Fluorobenzaldehyde, 97%
Thermo Scientific Chemicals
3-Fluorobenzaldehyde, 97%
CAS: 456-48-4 | C7H5FO | 124.114 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALFA11396.18
Price (MYR)
613.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
613.00
EA
Add to cart
Specifications
Chemical Name or Material3-Fluorobenzaldehyde
CAS456-48-4
Health Hazard 1H226-H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c