Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFA11379.30
Price (MYR)
827.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
827.00
EA
Add to cart
Specifications
Chemical Name or MaterialTris (hydroxymethyl)aminomethane hydrochloride
CAS1185-53-1
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point∼152°C (decomposition)
View more
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only
General References:
  1. Biological buffer.
  2. Lin, L.; Pan, Q. ZnFe2O4@ C/graphene nanocomposites as excellent anode materials for lithium batteries. J. Mater. Chem. A 2015, 3 (4), 1724-1729.
  3. Oishi, M. Enzyme-free and isothermal detection of microRNA based on click-chemical ligation-assisted hybridization coupled with hybridization chain reaction signal amplification. Anal. Bioanal. Chem. 2015, 407 (14), 4165-4172.