Sodium fumarate, 98%
Sodium fumarate, 98%
Sodium fumarate, 98%
Sodium fumarate, 98%
Thermo Scientific Chemicals
Sodium fumarate, 98%
CAS: 17013-01-3 | C4H4Na2O4 | 162.052 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11276.22100 g
Catalog number ALFA11276.22
Price (MYR)
289.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
289.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium fumarate
CAS17013-01-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Harry-O’kuru, R. E.; Gordon, S. H.; Klokkenga, M. Bio-generation of succinic acid by fermentation of Physaria fendleri seed polysaccharides. Ind. Crops Prod. 2015, 77, 116-122.
  2. Staszak, K.; Woźniak, M.; Sottek, M.; Karaś, Z.; Prochaska, K. Removal of fumaric acid from simulated and real fermentation broth. J. Chem. Technol. Biotechnol. 2015, 90 (3), 432-440.