L-Alanine ethyl ester hydrochloride, 98+%
L-Alanine ethyl ester hydrochloride, 98+%
L-Alanine ethyl ester hydrochloride, 98+%
L-Alanine ethyl ester hydrochloride, 98+%
Thermo Scientific Chemicals
L-Alanine ethyl ester hydrochloride, 98+%
CAS: 1115-59-9 | C5H12ClNO2 | 153.61 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11271.065 g
Catalog number ALFA11271.06
Price (MYR)
368.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
368.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-Alanine ethyl ester hydrochloride
Melting Point74°C to 78°C
CAS1115-59-9
Recommended StorageAmbient temperatures
EINECS Number214-225-9