D-(-)-Tartaric acid, 99%
D-(-)-Tartaric acid, 99%
D-(-)-Tartaric acid, 99%
D-(-)-Tartaric acid, 99%
Thermo Scientific Chemicals
D-(-)-Tartaric acid, 99%
CAS: 147-71-7 | C4H6O6 | 150.09 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11264.36500 g
Catalog number ALFA11264.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD- (-)-Tartaric acid
pH1.6
CAS147-71-7
Melting Point166°C to 170°C
Health Hazard 1H318-H500
View more
D-(-)-Tartaric acid is used as a resolving agent in organic synthesis. It is used as a precursor for the preparation of its ester derivatives like D-tartaric acid diethyl ester, D-tartaric acid dimethyl ester and D-tartaric acid diiso-propyl ester. It finds application in the synthesis of chiral aziridine derivative, a common intermediate for the preparation of hydroxyethylamine class HIV protease inhibitors viz. as saquinavir, amprenavir and nelfinavir. It is widely used in the food industry as a beer foaming agent, for food acidity regulations and as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-(-)-Tartaric acid is used as a resolving agent in organic synthesis. It is used as a precursor for the preparation of its ester derivatives like D-tartaric acid diethyl ester, D-tartaric acid dimethyl ester and D-tartaric acid diiso-propyl ester. It finds application in the synthesis of chiral aziridine derivative, a common intermediate for the preparation of hydroxyethylamine class HIV protease inhibitors viz. as saquinavir, amprenavir and nelfinavir. It is widely used in the food industry as a beer foaming agent, for food acidity regulations and as a flavoring agent.
Solubility
Soluble in water and alcohol.
Notes
Light sensitive. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only
General References:
  1. Mahapatra, M.; Tysoe, W. T. Structure and decomposition pathways of D-(-)-tartaric acid on Pd(111). Surface Science 2014, 629, 132-138.
  2. Hu, G.; Zhang, H.; Miao, H.; Wang, J.; Xu, Y. Synthesis, structures and properties of two new chiral rare earth-organic frameworks constructed by l/d-tartaric acid. J. Solid State Chem. 2015, 229, 208-212.