Ammonium iron(III) citrate, brown
Ammonium iron(III) citrate, brown
Ammonium iron(III) citrate, brown
Ammonium iron(III) citrate, brown
Thermo Scientific Chemicals
Ammonium iron(III) citrate, brown
CAS: 1185-57-5 | C6H11FeNO7+3 | 264.999 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11199.30250 g
Catalog number ALFA11199.30
Price (MYR)
270.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
270.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium iron (III) citrate
CAS1185-57-5
ColorBrown
Recommended StorageAmbient temperatures
RTECS NumberGE7540000
View more
Ammonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ammonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.
Solubility
Soluble in water.1200 g/L (20°C)
Notes
Hygroscopic. Desiccate at room temperature. Store in the dark. Store under inert gas.
RUO – Research Use Only
General References:
  1. Malcolm P. N. Brown J. J. Hahn, P. F. Stillman, A. E. Li, K. C.P., Kawamura Y. Tanaka, T. Noel J. K. Molony B. A. Johnson M. F. and Hildebolt C. F. The clinical value of ferric ammonium citrate: A positive oral contrast agent for T1-weighted MR imaging of the upper abdomen. J. Magn. Reson. Imaging. 2000, 702-707.
  2. D Richardson and E.baker. Two mechanisms of iron uptake from transferrin by melanoma cells. The effect of desferrioxamine and ferric ammonium citrate. Journal of biological chemistry. 1992, 267, 13972-13979.