Ethyl dodecanoate, 98%
Ethyl dodecanoate, 98%
Ethyl dodecanoate, 98%
Ethyl dodecanoate, 98%
Thermo Scientific Chemicals
Ethyl dodecanoate, 98%
CAS: 106-33-2 | C14H28O2 | 228.38 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11188.AE100 mL
Catalog number ALFA11188.AE
Price (MYR)
323.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
323.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthyl dodecanoate
CAS106-33-2
Melting Point-2°C
Recommended StorageAmbient temperatures
Density0.862