Vanillin, 99%
Vanillin, 99%
Vanillin, 99%
Thermo Scientific Chemicals
Vanillin, 99%
CAS: 121-33-5 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number ALFA11169.36
Price (MYR)
783.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
783.00
EA
Add to cart
Specifications
Chemical Name or MaterialVanillin
CAS121-33-5
Health Hazard 1H319-H500
Health Hazard 2GHS H Statement
H302-H332
Health Hazard 3P264b-P280i-P305+P351+P338
View more
Vanillin is a flavoring agent used in sweet foods, ice cream and chocolate. It is used in perfumes as well as to control the unpleasant odors in medicines. It acts as a chemical intermediate in the preparation of pharmaceuticals. In thin layer chromatography, it is used as a general-purpose stain for developing TLC plates to enable in visualizing components of a reaction mixture. Similarly, vanillin hydro chloride staining can be used to visualize the localization of tannins in cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Vanillin is a flavoring agent used in sweet foods, ice cream and chocolate. It is used in perfumes as well as to control the unpleasant odors in medicines. It acts as a chemical intermediate in the preparation of pharmaceuticals. In thin layer chromatography, it is used as a general-purpose stain for developing TLC plates to enable in visualizing components of a reaction mixture. Similarly, vanillin hydro chloride staining can be used to visualize the localization of tannins in cells.
Solubility
Soluble in water, diethyl ether, acetone, methanol, benzene, petroleum ether, chloroform, carbon disulfide, glacial acetic acid, pyridine and in aqueous solutions of alkali hydroxides.
Notes
Moisture and light sensitive. Incompatible with oxidizing agents, alkalis and reducing agents.
RUO – Research Use Only
General References:
  1. Tomadoni, B.; Cassani, L.; Moreira, M. R.; Ponce, A. Efficacy of vanillin and geraniol in reducing Escherichia coli O157: H7 on strawberry juice. LWT Food Sci. Technol. 2015, 64 (2), 554-557.
  2. Di Paola, A.; Bellardita, M.; Megna, B.; Parrino, F.; Palmisano, L. Photocatalytic oxidation of trans-ferulic acid to vanillin on TiO2 and WO3-loaded TiO2 catalysts. Catal. Today 2015, 252, 195-200.