Potassium phthalimide, 98+%
Potassium phthalimide, 98+%
Potassium phthalimide, 98+%
Potassium phthalimide, 98+%
Thermo Scientific Chemicals
Potassium phthalimide, 98+%
CAS: 1074-82-4 | C8H4KNO2 | 185.22 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11134.22100 g
Catalog number ALFA11134.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium phthalimide
CAS1074-82-4
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Potassium phthalimide is used as an intermediate in the synthesis of N-alkylated phthalimides, which is involved in the preparation of primary amines (Gabriel synthesis) by the hydrolysis reaction. It is also used as an intermediate for synthetic indigo, pigments, dyes and pharmaceuticals. Further, it is employed as an organocatalyst for the cyanosilylation of various carbonyl compounds under extremely mild conditions. In addition to this, it serves as a reagent for the transformation of allyl- and alkyl halides into protected primary amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium phthalimide is used as an intermediate in the synthesis of N-alkylated phthalimides, which is involved in the preparation of primary amines (Gabriel synthesis) by the hydrolysis reaction. It is also used as an intermediate for synthetic indigo, pigments, dyes and pharmaceuticals. Further, it is employed as an organocatalyst for the cyanosilylation of various carbonyl compounds under extremely mild conditions. In addition to this, it serves as a reagent for the transformation of allyl- and alkyl halides into protected primary amines.
Solubility
Soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. For an example of the classical Gabriel synthesis of primary amines from an alkyl halide by alkylation of potassium phthalimide followed by hydrolysis, see: Org. Synth. Coll., 2, 25 (1943). For use of the Gabriel reaction on Merrifield resin as the first step of a synthesis of a polymer-supported carbodiimide, see: Org. Synth. Coll., 6, 951 (1988). The phthalimide alkylation is often carried out in DMF. A liquid-liquid phase-transfer method using (n-Hexadecyl) tri-n-butyl phosphonium bromide, L01335, as catalyst also gives excellent results: Synthesis, 389 (1976).
  2. The standard method for cleavage of the N-substituted phthalimide is with hydrazine hydrate, effective under milder conditions than acid or base hydrolysis. Other hydrazines and primary amines have also been successful, e.g. methylhydrazine or N,N-dimethylpropanediamine: Org. Synth. Coll., 9, 13, 16 (1998). Another technique involves borohydride reduction to the hydroxy amide, and liberation of the amine with acetic acid, with cyclization to phthalide: Tetrahedron Lett., 25, 2093 (1984). For suggested improvements to the cleavage conditions in the Gabriel synthesis, see: J. Org. Chem., 61, 8063 (1996). Review of the Gabriel reaction: Angew. Chem. Int. Ed., 7, 919 (1968).
  3. For an alternative approach to the formation of primary amines, see Dibenzyl amine, A11554.
  4. Kiyokawa, K.; Kosaka, T.; Kojima, T.; Minakata, S. Synthesis and Structure of Hypervalent Iodine(III) Reagents Containing Phthalimidate and Application to Oxidative Amination Reactions. Angew. Chem. Int. Ed. 2015, 127 (46), 13923-13927.
  5. Fesenko, A. A.; Trafimova, L. A.; Albov, D. V.; Shutalev, A. D. Nucleophile-dependent diastereoselectivity in the ring expansion of pyrimidines to give 1,3-diazepines. Tetrahedron Lett. 2015, 56 (11), 1317-1321.