5-Chlorouracil, 98%
5-Chlorouracil, 98%
5-Chlorouracil, 98%
Thermo Scientific Chemicals
5-Chlorouracil, 98%
CAS: 1820-81-1 | C4H3ClN2O2 | 146.53 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11084.1425 g
Catalog number ALFA11084.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Chlorouracil
CAS1820-81-1
Melting Point>300°C
Recommended StorageAmbient temperatures
RTECS NumberYQ9410000