Sodium butyrate, 98+%
Sodium butyrate, 98+%
Sodium butyrate, 98+%
Sodium butyrate, 98+%
Thermo Scientific Chemicals
Sodium butyrate, 98+%
CAS: 156-54-7 | C4H7NaO2 | 110.09 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11079.22100 g
Catalog number ALFA11079.22
Price (MYR)
373.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
373.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium butyrate
CAS156-54-7
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point>300°C
View more
Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.
Solubility
Soluble in water, dimethyl sulfoxide and ethanol. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Cavini, S.; Iraira, S.; Siurana, A.; Foskolos, A.; Ferret, A.; Calsamiglia, S. Effect of sodium butyrate administered in the concentrate on rumen development and productive performance of lambs in intensive production system during the suckling and the fattening periods. Small Rumin. Res. 2015, 123 (2-3), 212-217.
  2. Xu, H.; McCoy, A.; Li, J.; Zhao, Y.; Ghishan, F. K. Sodium butyrate stimulates NHE8 expression via its role on activating NHE8 basal promoter activity. Am. J. Physiol. Gastrointest. Liver Physiol. 2015, 309 (6), G500-G505.