Xanthine, 99%
Xanthine, 99%
Xanthine, 99%
Thermo Scientific Chemicals
Xanthine, 99%
CAS: 69-89-6 | C5H4N4O2 | 152.113 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11077.1425 g
Catalog number ALFA11077.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialXanthine
CAS69-89-6
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point>300°C
View more
Xanthine and xanthine oxidase system can be used to produce superoxide radicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Xanthine and xanthine oxidase system can be used to produce superoxide radicals.
Solubility
Soluble in water(0.067g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Su-Shu Pan and Nicholas R. Bachur. Xanthine Oxidase Catalyzed Reductive Cleavage of Anthracycline Antibiotics and Free Radical Formation. Molecular Pharmacology. 1980, 17 (1),95-99.
  2. Mario Thevis,; Georg Opfermann,; Oliver Krug,; Wilhelm Schänzer. Electrospray ionization mass spectrometric characterization and quantitation of xanthine derivatives using isotopically labelled analogues: an application for equine doping control analysis. Rapid Communications in Mass Spectrometry. 2004, 18 (14),1553-1560.