Formamide, 99%
Formamide, 99%
Formamide, 99%
Thermo Scientific Chemicals
Formamide, 99%
CAS: 75-12-7 | CH3NO | 45.04 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11076.36500 g
Catalog number ALFA11076.36
Price (MYR)
391.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
391.00
EA
Add to cart
Specifications
Chemical Name or MaterialFormamide
Melting Point2°C
CAS75-12-7
Health Hazard 1H351-H360FD-H373
Health Hazard 2GHS H Statement
H360;
May damage fertility or the unborn child.
View more
Formamide is used for cryopreservation of tissues and organs and in hybridization buffers. It acts as an RNA stabiliser in gel electrophoresis by deionizing RNA. It is used as an alternative solvent for the electrostatic self-assembly of polymer nanofilms. It is used in the preparation of brain sections of mice for BrdU immunohistochemistry and fish analysis of HeLa cells. It essentially plays a vital role in crystallization of pharmaceuticals, in soil stabilization, as a solvent in inks, as a component of liquid fertilizers, as a heat-resistant coatings and in some personal care products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Formamide is used for cryopreservation of tissues and organs and in hybridization buffers. It acts as an RNA stabiliser in gel electrophoresis by deionizing RNA. It is used as an alternative solvent for the electrostatic self-assembly of polymer nanofilms. It is used in the preparation of brain sections of mice for BrdU immunohistochemistry and fish analysis of HeLa cells. It essentially plays a vital role in crystallization of pharmaceuticals, in soil stabilization, as a solvent in inks, as a component of liquid fertilizers, as a heat-resistant coatings and in some personal care products.
Solubility
It is miscible with water.
Notes
Incompatible with bases, oxidizing agents, hydrogen peroxide, iodine, pyridine and sulphur oxides.
RUO – Research Use Only
General References:
  1. An, H.; Liu, G.; Rob Atkin, R.; Craig, V. S. J. Surface Nanobubbles in Nonaqueous Media: Looking for Nanobubbles in DMSO, Formamide, Propylene Carbonate, Ethylammonium Nitrate, and Propylammonium Nitrate. ACS Nano 2015, 9 (7), 7596-7607.
  2. Yang, Z.; Price, N. E.; Johnson, K. M.; Gates, K. S. Characterization of Interstrand DNA-DNA Cross-Links Derived from Abasic Sites Using Bacteriophage ϕ29 DNA Polymerase. Biochemistry 2015, 54 (27), 4259-4266.