Diethylenetriamine, 99%
Diethylenetriamine, 99%
Diethylenetriamine, 99%
Diethylenetriamine, 99%
Thermo Scientific Chemicals
Diethylenetriamine, 99%
CAS: 111-40-0 | C4H13N3 | 103.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11033.AE100 mL
Catalog number ALFA11033.AE
Price (MYR)
315.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
315.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiethylenetriamine
CAS111-40-0
Health Hazard 1H227-H302+H312-H314-H317-H330-H335
Health Hazard 2GHS H Statement
H314-H302-H312-H317
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Health Hazard 3P210-P235-P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
View more
Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.
Solubility
Miscible with water and ethanol. Immiscible with ether.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and copper.
RUO – Research Use Only
General References:
  1. Galhoum, A. A.; Mahfouz, M. G.; Rehem, S. T. A.; Gomaa, N. A.; Atia, A. A.; Vincent, T.; Guibal, E. Diethylenetriamine-functionalized chitosan magnetic nano-based particles for the sorption of rare earth metal ions [Nd(III), Dy(III) and Yb(III)]. Cellulose 2015, 22 (4), 2589-2605.
  2. Koizumi, Y.; Hirooka, M.; Ochi, H.; Tokumoto, Y.; Takechi, M.; Hiraoka, A.; Ikeda, Y.; Kumagi, T.; Matsuura, B. Characterization of the biliary tract by virtual ultrasonography constructed by gadolinium ethoxybenzyl diethylenetriamine pentaacetic acid-enhanced magnetic resonance imaging. J Med Ultrason 2015, 42 (2), 185-193.