Methyl octanoate, 98+%
Methyl octanoate, 98+%
Methyl octanoate, 98+%
Methyl octanoate, 98+%
Thermo Scientific Chemicals
Methyl octanoate, 98+%
CAS: 111-11-5 | C9H18O2 | 158.241 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFA10991.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl octanoate
CAS111-11-5
Health Hazard 1H227
Health Hazard 3P210-P280-P370+P378q-P501c
Melting Point-40°C
View more
Methyl octanoate-ethanol mixtures constitute the biodiesel-bioethanol surrogate fuel and kinetics of its oxidation has been studied experimentally in a jet-stirred reactor. Deoxygenation of methyl octanoate over alumina-supported Pt has been studied in both the vapor phase in a flow reactor and in the liquid phase in a semi-batch reactor. It is also used to make other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl octanoate-ethanol mixtures constitute the biodiesel-bioethanol surrogate fuel and kinetics of its oxidation has been studied experimentally in a jet-stirred reactor. Deoxygenation of methyl octanoate over alumina-supported Pt has been studied in both the vapor phase in a flow reactor and in the liquid phase in a semi-batch reactor. It is also used to make other chemicals.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. C.Togbe; J.B.May-Carle; G.Dayma; P.Dagaut. Chemical kinetic study of the oxidation of a biodiesel-bioethanol surrogate fuel: methyl octanoate-ethanol mixtures. Journal of Physical Chemistry A: Molecules, Spectroscopy, Kinetics, Environment and General Theory. 2010, 114,(11), 3896-3908.
  2. Phuong T.Do; Martina Chiappero; Lance L.Lobban; Daniel E.Resasco. Catalytic Deoxygenation of Methyl-Octanoate and Methyl-Stearate on Pt/Al2O3. Catalysis Letters. 2009, 130,(1-2), 9-18.