N,N-Dimethylacetamide, 99%
N,N-Dimethylacetamide, 99%
N,N-Dimethylacetamide, 99%
N,N-Dimethylacetamide, 99%
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number ALFA10924.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialN,N-Dimethylacetamide
CAS127-19-5
Health Hazard 1H227-H312+H332-H360D
Health Hazard 2GHS H Statement
H360-H312-H332-H227
May damage fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Health Hazard 3P201-P202-P210-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P370+P378q-P501c
View more
N,N-Dimethylacetamide is used as a polar solvent in organic synthesis as well as in reactions involving strong bases such as sodium hydroxide. It is also used as a solvent for fibers like polyacrylonitrile, spandex, as an excipient in drugs viz. vumon, busulfex and in the adhesive industry. It finds application as a reaction medium in the production of pharmaceuticals, agrochemicals, dyes and plasticizers. It is widely used in polymer industry due to its high solving power for high molecular-weight polymers and synthetic resins. It plays an important role as a catalyst in various reactions viz. cyclization, halogenation, cynidation, alkylation and dehydrogenation and increases the yield of main products. It is also used as an extracting agent for oil and gases, in paint removers, in the production of photo-resist stripping compounds and as booster solvent in coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Dimethylacetamide is used as a polar solvent in organic synthesis as well as in reactions involving strong bases such as sodium hydroxide. It is also used as a solvent for fibers like polyacrylonitrile, spandex, as an excipient in drugs viz. vumon, busulfex and in the adhesive industry. It finds application as a reaction medium in the production of pharmaceuticals, agrochemicals, dyes and plasticizers. It is widely used in polymer industry due to its high solving power for high molecular-weight polymers and synthetic resins. It plays an important role as a catalyst in various reactions viz. cyclization, halogenation, cynidation, alkylation and dehydrogenation and increases the yield of main products. It is also used as an extracting agent for oil and gases, in paint removers, in the production of photo-resist stripping compounds and as booster solvent in coatings.
Solubility
Miscible with water, benzene, alcohol, acetone, ether, esters and ketones.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Dipolar aprotic solvent, cf N,N-Dimethyl formamide, A13547, Dimethyl sulfoxide, A13280, 1-Methyl-2-pyrrolidinone, A12260.
  2. See also Sodium hydride, 13431, for warning.
  3. Useful solvent in halex fluorinations of chloroarenes; compare Sulfolane, A13466.
  4. Undergoes Vilsmeier-type reactions (compare DMF), with reactive substrates, introducing an acetyl group: Chem. Ber., 97, 616 (1964).
  5. Reacts with alkyl and aryl Grignard reagents to give methyl ketones in good yields: Synthesis, 228 (1984).
  6. In the presence of triflic anhydride and 2,4,6-collidine, forms a highly-reactive keteniminium triflate, which undergoes a [2+2] cycloaddition reaction with terminal alkenes to give, after hydrolysis a 3-alkylcyclobutanone: J. Am. Chem. Soc., 107, 2192 (1985); Org. Synth. Coll., 8, 306 (1993):
  7. Messaadi, A.; Salhi, H.; Das, D.; Alzamil, N. O.; Alkhaldi, M. A.; Ouerfelli, N.; Hamzaoui, A. H. A novel approach to discuss the viscosity Arrhenius behaviour and to derive the partial molar properties in binary mixtures of N, N-dimethylacetamide with 2-methoxyethanol in the temperature interval (from 298.15 to 318.15) K. Phys. Chem. Liq. 2015, 53 (4), 506-517.
  8. Shokouhi, M.; Farahani, H.; Jenab, M. H.; Jalili, A. H. Solubility of Hydrogen Sulfide in N-Methylacetamide and N, N-Dimethylacetamide: Experimental Measurement and Modeling. J. Chem. Eng. Data 2015, 60 (3), 499-508.