N-(2-Hydroxyethyl)ethylenediamine, 99%
N-(2-Hydroxyethyl)ethylenediamine, 99%
N-(2-Hydroxyethyl)ethylenediamine, 99%
N-(2-Hydroxyethyl)ethylenediamine, 99%
Thermo Scientific Chemicals
N-(2-Hydroxyethyl)ethylenediamine, 99%
CAS: 111-41-1 | C4H12N2O | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA10833.36
Price (MYR)
382.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
382.00
EA
Add to cart
Specifications
Chemical Name or MaterialN- (2-Hydroxyethyl)ethylenediamine
CAS111-41-1
Health Hazard 1H314-H317-H360Df
Health Hazard 2GHS H Statement
H360-H314-H318-H317
May damage fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
View more
It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.
Solubility
Fully miscible in water.
Notes
Air Sensitive & Hygroscopic. Store under inert gas. Keep away from incompatibles.
RUO – Research Use Only
General References:
  1. Hassan Ali Zamani.; Mohammad Taghi Hamed-Mosavian.;Elham Hamidfar.; Mohammad Reza Ganjali.; Parviz Norouzi. A novel iron (III)-PVC membrane potentiomeric sensor based on N-(2-hydroxyethyl)ethylenediamine-N,N',N″-triacetic acid. Materials Science and Engineering: C. 2008, 28, (8), 1551-1555.
  2. Hiroshi Ogino.; Mamoru Shimoi.; Yoko Saito. Structural identification of the reactive vanadium(III) intermediate formed in the electron-transfer reactions of [N'-(2-hydroxyethyl)ethylenediamine-N,N,N'-triacetato]aquavanadium(III) complex ([V(hedtra)(H2O)]) with halopentaamminecobalt(III) complexes: x-ray crystal structures of [V(hedtra)(H2O)].cntdot.2H2O and K[VO(hedtra)].cntdot.H2O. norg. Chem 1989, 28, (18), 3596-3600.