2,4-Difluoro-1-iodobenzene, 99%
2,4-Difluoro-1-iodobenzene, 99%
2,4-Difluoro-1-iodobenzene, 99%
2,4-Difluoro-1-iodobenzene, 99%
Thermo Scientific Chemicals
2,4-Difluoro-1-iodobenzene, 99%
CAS: 2265-93-2 | C6H3F2I | 239.99 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10798.0910 g
Catalog number ALFA10798.09
Price (MYR)
287.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
287.00
EA
Add to cart
Specifications
Chemical Name or Material2,4-Difluoro-1-iodobenzene
CAS2265-93-2
Health Hazard 1H227-H302-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c