Adenosine, 99%
Adenosine, 99%
Adenosine, 99%
Thermo Scientific Chemicals
Adenosine, 99%
CAS: 58-61-7 | C10H13N5O4 | 267.25 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10781.0910 g
Catalog number ALFA10781.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAdenosine
CAS58-61-7
Melting Point234°C to 237°C
Recommended StorageKeep cold
RTECS NumberAU7175000
View more
Adenosine serves as a nucleotide. It also plays an important role in the regulation of blood flow to various organs through vasodilation. It is actively involved in biochemical processes, such as energy transfer as adenosine triphosphate and adenosine diphosphate. Further, it is used in medication specifically as an antiarrhythmic agent to treat a number of supraventricular tachycardia. It is used to promote thickening of hair for people with thinning hair.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water, ammonium hydroxide and dimethyl sulfoxide. Insoluble in ethanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Lebon, G.; Edwards, P. C.; Leslie, A. G.; Tate, C. G. Molecular Determinants of CGS21680 Binding to the Human Adenosine A2A Receptor. Mol. Pharmacol. 2015, 87 (6), 907-915.
  2. Carlsson, M.; Jögi, J.; Bloch, K. M.; Hedén, B.; Ekelund, U.; Ståhlberg, F.; Arheden, H. Submaximal adenosine-induced coronary hyperaemia with 12 h caffeine abstinence: implications for clinical adenosine perfusion imaging tests. Clin. Physiol. Funct. Imaging 2015, 35 (1), 49-56.