Agar powder
Agar powder
Agar powder
Thermo Scientific Chemicals
Agar powder
CAS: 9002-18-0 | C14H24O9 | 336.337 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA10752.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAgar
Physical FormPowder
CAS9002-18-0
Recommended StorageAmbient temperatures
RTECS NumberAW7950000
View more
Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.
Solubility
Soluble in water at temperatures between 950C and 1000 C. Insoluble in cold water.
Notes
Prolonged exposure to high temperatures can degrade Agar powder.
RUO – Research Use Only
General References:
  1. For gel preparation.
  2. Chi, W. J.; Chang, Y. K.; Hong, S. K. Agar degradation by microorganisms and agar-degrading enzymes. Appl. Microbiol. Biotechnol. 2012, 94 (4), 917-30.
  3. Laurienzo, P. Marine polysaccharides in pharmaceutical applications: An overview. Mar. Drugs. 2010, 8 (9), 2435-2465.