2,3,5-Trimethylphenol, 98+%
2,3,5-Trimethylphenol, 98+%
2,3,5-Trimethylphenol, 98+%
2,3,5-Trimethylphenol, 98+%
Thermo Scientific Chemicals
2,3,5-Trimethylphenol, 98+%
CAS: 697-82-5 | C9H12O | 136.194 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10745.22100 g
Catalog number ALFA10745.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2,3,5-Trimethylphenol
CAS697-82-5
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c