Heptafluorobutyric anhydride, 98%
Heptafluorobutyric anhydride, 98%
Heptafluorobutyric anhydride, 98%
Heptafluorobutyric anhydride, 98%
Thermo Scientific Chemicals
Heptafluorobutyric anhydride, 98%
CAS: 336-59-4 | C8F14O3 | 410.06 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10744.065 g
Catalog number ALFA10744.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHeptafluorobutyric anhydride
CAS336-59-4
Health Hazard 1H314
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Heptafluorobutyric anhydride is used as a derivatization reagent for gas chromatography - mass mass spectrometry analysis of the stereoisomer content of synthetic peptides. It is a GC derivatizing agent used to prepare volatile derivatives of amino acids and other biologically active compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Heptafluorobutyric anhydride is used as a derivatization reagent for gas chromatography - mass mass spectrometry analysis of the stereoisomer content of synthetic peptides. It is a GC derivatizing agent used to prepare volatile derivatives of amino acids and other biologically active compounds.
Solubility
Miscible with halogenated solvents. Immiscible with polar solvents.
Notes
Incompatible with moist air, water, strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only
General References:
  1. Cappelle, D.; Neels, H.; Yegles, M.; Paulus, J.; van Nuijs, A. L.; Covaci, A.; Crunelle, C. L. Gas chromatographic determination of ethyl glucuronide in hair: Comparison between tandem mass spectrometry and single quadrupole mass spectrometry. Forensic Sci. Int. 2015, 249, 20-24
  2. Nikolaou, P.; Papoutsis, I.; Spiliopoulou, C.; Voudris, C.; Athanaselis, S. A fully validated method for the determination of lacosamide in human plasma using gas chromatography with mass spectrometry: Application for therapeutic drug monitoring. J. Sep. Sci. 2015, 38, (2), 260-266.